Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE10A Mouse anti-Human, Clone: OTI1B2, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500635S

Description
Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate.Specifications
PDE10A | |
Monoclonal | |
0.6 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y233 | |
PDE10A | |
Full-length protein expressed in 293T cell transfected with human PDE10A expression vector | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1B2 | |
Unconjugated | |
PDE10A | |
ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10846 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction