Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA131145
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA131145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA131145 Supplier Invitrogen™ Supplier No. PA131145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: For western blot - Rat brain and pancreas.

Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE10A
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation Tris glycine with 0.5% BSA, 30% glycerol and 0.02% Sodium azide; pH 7.4-7.9
Gene PDE10A
Gene Accession No. Q8CA95, Q9QYJ6, Q9Y233
Gene Alias ADSD2; cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A; dJ416F21.1 (phosphodiesterase 10A); dual-substrate cAMP and cGMP, cyclic nucleotide phosphodiesterase; FLJ11894; FLJ25677; HSPDE10A; IOLOD; LINC00473; PDE10A; PDE10A19; PDE10A2 brain-specific isoform; Pde10a3; phosphodiesterase 10A; phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A7 (PDE10A7); testis-specific phosphodiesterase PDE10A2
Gene Symbols PDE10A
Host Species Rabbit
Immunogen Synthetic peptide (Human) taken against a common sequence near the 250-325 amino acids downstream of N-terminal region of PDE10A. Synthetic MAP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10846, 23984, 63885
Target Species Human, Mouse, Monkey, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.