Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116660 Supplier Invitrogen™ Supplier No. PA5116660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Mouse testis, Mouse liver, Rat brain, Rat testis, Rat liver Immunogen sequence: NNAFQAKSGS ALAQLYGTSA TLEHHHFNHA VMILQSEGHN IFANLSSKEY SDLMQLLKQS ILATDLTLYF ERRTEFFELV SKGEYDWNIK NHRDIFRSML MTACDLGAVT KPWEISRQVA ELVTSEFFEQ GDRERLELKL TPSAIFDRNR KDELPRLQLE WIDSICMPLY QALVKVNVKL KPMLDSVATN RSKWEELHQK RLLASTASSS PASVMVAKED RN.

The 3',5'-cyclic nucleotides cAMP and cGMP function as second messengers in a wide variety of signal transduction pathways. 3',5'-cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and cGMP to the corresponding 5'-monophosphates and provide a mechanism to downregulate cAMP and cGMP signaling. This gene encodes a member of the PDE protein superfamily. Mutations in this gene are a cause of Cushing disease and adrenocortical hyperplasia. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE11A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PDE11A
Gene Accession No. P0C1Q2, Q8VID6
Gene Alias 6330414F14Rik; A630086N24Rik; cAMP and cGMP cyclic nucleotide phosphodiesterase 11A; cAMP and cGMP phosphodiesterase 11A; Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A; FLJ23693; Gm350; MGC133355; MGC133356; Pde11a; PDE11A1; phosphodiesterase 11A; PPNAD2
Gene Symbols PDE11A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 712-933 of human PDE11A (NP_0586493).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140928, 241489
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.