Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544028 Supplier Invitrogen™ Supplier No. PA544028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FGLEGVFRIK QHEGLATFYR KSKFSLLSQH DISFYEALES DPLHKELLEK Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 79%.

PDE12 is an enzyme that cleaves 2',5'-phosphodiester bond linking adenosines of the 5'-triphosphorylated oligoadenylates, triphosphorylated oligoadenylates referred as 2-5A modulates the 2-5A system. This enzyme degraded triphosphorylated 2-5A to produce AMP and ATP. It also cleaves 3',5'-phosphodiester bond of oligoadenylates and plays a role as a negative regulator of the 2-5A system that is one of the major pathways for antiviral and antitumor functions induced by interferons (IFNs). Suppression of this enzyme induces reduction of viral replication in Hela cells, thus counteracting the antiviral pathway probably by inhibiting the 2-5A system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDE12
Gene Accession No. Q6L8Q7
Gene Alias 2',5'-phosphodiesterase 12; 2',5'-phosphodiesterase 12-like protein; 2'-PDE; 2-PDE; 2'-phosphodiesterase; E430028B21Rik; fc58c05; fi42f03; LOC558284 protein; LOW QUALITY PROTEIN: 2',5'-phosphodiesterase 12; LRRGT00074; Mitochondrial deadenylase; PDE12; phosphodiesterase 12; RGD1310975; wu:fc58c05; wu:fi42f03; zgc:136374
Gene Symbols PDE12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PDE12 (aa 368-417).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201626
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.