Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE1A Antibody (OTI7D5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273277APC
Description
PDE1A Monoclonal antibody specifically detects PDE1A in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
PDE1A | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
5136 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI7D5 | |
Western Blot, Immunohistochemistry | |
3'-5' cyclic nucleotide phosphodiesterase, 61 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1A, calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, calmodulin-dependent phosphodiesterase, Cam-PDE 1A, EC 3.1.4, EC 3.1.4.17, hCam-1, HCAM1, HSPDE1A, MGC26303, phosphodiesterase 1A, calmodulin-dependent, phosphodiesterase-1A | |
Human recombinant protein fragment corresponding to amino acids 259-545 of human PDE1A (NP_005010) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction