Learn More
Invitrogen™ PDE1A Monoclonal Antibody (OTI7D5)

Description
PDE1A Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PDE1A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI7D5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PDE1A |
| Gene Accession No. | P54750 |
| Gene Alias | 3'-RACE clone 8 phosphodiesterase 1A; 3-RACE clone 8 phosphodiesterase 1A; 3'-RACEclone8phosphodiesterase1A; 3-RACEclone8phosphodiesterase1A; 61 kDa Cam-PDE; AI987702; AW125737; calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; cam-PDE 1A; CAM-PDE-1A; HCAM1; hCam-1; HS MGC26303; HSPDE1A; MGC26303; Pde1a; phosphodiesterase 1A; phosphodiesterase 1A, calmodulin-dependent; phosphodiesterase-1A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.