Learn More
PDE1A Mouse anti-Human, Clone: OTI7D5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803629

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclic nucleotide phosphodiesterases play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Cathat are activated by calmodulin in the presence of Ca.Specifications
PDE1A | |
Monoclonal | |
Unconjugated | |
PDE1A | |
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A | |
Mouse | |
Affinity Chromatography | |
RUO | |
5136 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7D5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P54750 | |
PDE1A | |
Human recombit protein fragment corresponding to amino acids 259-545 of human PDE1A produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.