Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582794 Supplier Invitrogen™ Supplier No. PA582794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNLVIEMVLS TDMSGHFQQI KNIRNSLQQP EGIDRAKTMS LILHAADISH PAKSWKLHYR WTMALMEEFF LQGDKEAE Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 97%.

Cyclic nucleotide phosphodiesterases (PDEs) play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Ca(2+)/calmodulin (see CALM1)that are activated by calmodulin in the presence of Ca(2+) (Michibata et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDE1A
Gene Accession No. P54750
Gene Alias 3'-RACE clone 8 phosphodiesterase 1A; 3-RACE clone 8 phosphodiesterase 1A; 3'-RACEclone8phosphodiesterase1A; 3-RACEclone8phosphodiesterase1A; 61 kDa Cam-PDE; AI987702; AW125737; calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; cam-PDE 1A; CAM-PDE-1A; HCAM1; hCam-1; HS MGC26303; HSPDE1A; MGC26303; Pde1a; phosphodiesterase 1A; phosphodiesterase 1A, calmodulin-dependent; phosphodiesterase-1A
Gene Symbols PDE1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDE1A. Recombinant protein control fragment (Product #RP-89902).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.