Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE1B Monoclonal Antibody (OTI1D6), TrueMAB™, OriGene
SDP

Catalog No. 502540974
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0974 30 μL
50-167-4256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0974 Supplier OriGene Supplier No. TA503336S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE1B Monoclonal antibody specifically detects PDE1B in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PDE1B
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1D6
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Nudt9
Gene Accession No. Q9BW91
Gene Alias 63 a Cam PDE; 63 a Cam-PDE; 63 kDa Cam-PDE; anti-oxidant protein 2; Aop2; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; calmodulin-stimulated phosphodiesterase PDE1B1; Cam PDE 1B; Cam PDE1B; cam-PDE 1B; cyclic nucleotide phosphodiesterase (CaM-PDE); EGK_03739; epididymis secretory sperm binding protein Li 79p; HEL-S-79p; PDE 1B; Pde1b; Pde1b1; PDES 1B; PDES1B; Phosphodiesterase; phosphodiesterase 1B; phosphodiesterase 1B, Ca2+calmodulin dependent; phosphodiesterase 1B, Ca2+-calmodulin dependent; phosphodiesterase 1B, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase 1B, calmodulin-dependent; phosphodiesterase 1B1 Ca2+-calmodulin dependent 63 kDa; phosphodiesterase 1B1, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase IB calmodulin-dependent; presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Host Species Mouse
Immunogen Full length human recombinant protein of human NUDT9 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53343
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.