Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE1B Monoclonal Antibody (OTI6B9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503325

Description
The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE1 subfamily. Members of the PDE1 family are calmodulin-dependent PDEs that are stimulated by a calcium-calmodulin complex. This PDE has dual-specificity for the second messengers, cAMP and cGMP, with a preference for cGMP as a substrate. cAMP and cGMP function as key regulators of many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
PDE1B | |
Monoclonal | |
0.95 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q01064, Q01066 | |
PDE1B | |
Full length human recombit protein of human PDE1B produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI6B9 | |
Unconjugated | |
PDE1B | |
HEL-S-79p, PDE1B1, PDES1B | |
Mouse | |
Affinity Chromatography | |
RUO | |
100856025, 29691, 5153 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction