Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553817 Supplier Invitrogen™ Supplier No. PA553817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TFSVLTDVAE KSVQPLADED SKSKNQPSFQ WRQPSLDVEV GDPNPDVVSF RSTWVKRIQE NKQKWKERAA SGITNQMSID ELSPCEEEAP PSPAEDEHNQ NGN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE1 subfamily. Members of the PDE1 family are calmodulin-dependent PDEs that are stimulated by a calcium-calmodulin complex. This PDE has dual-specificity for the second messengers, cAMP and cGMP, with a preference for cGMP as a substrate. cAMP and cGMP function as key regulators of many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pde1b
Gene Accession No. Q01064
Gene Alias 63 a Cam PDE; 63 a Cam-PDE; 63 kDa Cam-PDE; anti-oxidant protein 2; Aop2; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; calmodulin-stimulated phosphodiesterase PDE1B1; Cam PDE 1B; Cam PDE1B; cam-PDE 1B; cyclic nucleotide phosphodiesterase (CaM-PDE); EGK_03739; epididymis secretory sperm binding protein Li 79p; HEL-S-79p; PDE 1B; Pde1b; Pde1b1; PDES 1B; PDES1B; Phosphodiesterase; phosphodiesterase 1B; phosphodiesterase 1B, Ca2+calmodulin dependent; phosphodiesterase 1B, Ca2+-calmodulin dependent; phosphodiesterase 1B, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase 1B, calmodulin-dependent; phosphodiesterase 1B1 Ca2+-calmodulin dependent 63 kDa; phosphodiesterase 1B1, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase IB calmodulin-dependent; presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Gene Symbols Pde1b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDE1B. Recombinant protein control fragment (Product #RP-92038).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.