Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE1B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA598347

Catalog No. PIPA598347


Only null left
Add to Cart

Description

Description

The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE1 subfamily. Members of the PDE1 family are calmodulin-dependent PDEs that are stimulated by a calcium-calmodulin complex. This PDE has dual-specificity for the second messengers, cAMP and cGMP, with a preference for cGMP as a substrate. cAMP and cGMP function as key regulators of many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PDE1B
Polyclonal
Unconjugated
Pde1b
63 a Cam PDE, 63 a Cam-PDE, 63 kDa Cam-PDE, anti-oxidant protein 2, Aop2, Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B, calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, calmodulin-stimulated phosphodiesterase PDE1B1, Cam PDE 1B, Cam PDE1B, cam-PDE 1B, cyclic nucleotide phosphodiesterase (CaM-PDE), EGK_03739, epididymis secretory sperm binding protein Li 79p, HEL-S-79p, PDE 1B, Pde1b, Pde1b1, PDES 1B, PDES1B, Phosphodiesterase, phosphodiesterase 1B, phosphodiesterase 1B, Ca2+calmodulin dependent, phosphodiesterase 1B, Ca2+-calmodulin dependent, phosphodiesterase 1B, Ca2+-calmodulin dependent, 63 kDa, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase 1B1 Ca2+-calmodulin dependent 63 kDa, phosphodiesterase 1B1, Ca2+-calmodulin dependent, 63 kDa, phosphodiesterase IB calmodulin-dependent, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Rabbit
Antigen affinity chromatography
RUO
5153
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry, Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4
Q01064
Pde1b
Recombinant Human Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B protein (1-277AA)
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.