Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554497 Supplier Invitrogen™ Supplier No. PA554497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSQVGFIDFI VEPTFTVLTD MTEKIVSPLI DETSQTGGTG QRRSSLNSIS SSDAKRSGVK TSGSEGSAPI NNSVISVDYK SFKATWTE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze hydrolysis of the cyclic nucleotides cAMP and cGMP to the corresponding nucleoside 5-prime-monophosphates. Mammalian PDEs have been classified into several families based on their biochemical properties. Members of the PDE1 family, such as PDE1C, are calmodulin -dependent PDEs (CaM-PDEs) that are stimulated by a calcium-calmodulin complex (Repaske et al., 1992 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE1C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDE1C
Gene Accession No. Q14123
Gene Alias 3',5'-cyclic-AMP phosphodiesterase; 3',5'-cyclic-GMP phosphodiesterase; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1C; cam-PDE 1C; cyclic nucleotide phosphodiesterase 1 C; Hcam3; hCam-3; Human 3',5' cyclic nucleotide phosphodiesterase (HSPDE1C1A); PDE1C; phosphodiesterase 1C; phosphodiesterase 1C calmodulin-dependent (70kD); phosphodiesterase 1C, calmodulin-dependent 70kDa
Gene Symbols PDE1C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDE1C. Recombinant protein control fragment (Product #RP-92764).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.