Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114082 Supplier Invitrogen™ Supplier No. PA5114082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IDFIVEPTFT VLTDMTEKIV SPLIDETSQT GGTGQRRSSL NSISSSDAKR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze hydrolysis of the cyclic nucleotides cAMP and cGMP to the corresponding nucleoside 5-prime-monophosphates. Mammalian PDEs have been classified into several families based on their biochemical properties. Members of the PDE1 family, such as PDE1C, are calmodulin -dependent PDEs (CaM-PDEs) that are stimulated by a calcium-calmodulin complex (Repaske et al., 1992 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE1C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDE1C
Gene Accession No. Q14123
Gene Alias 3',5'-cyclic-AMP phosphodiesterase; 3',5'-cyclic-GMP phosphodiesterase; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1C; cam-PDE 1C; cyclic nucleotide phosphodiesterase 1 C; Hcam3; hCam-3; Human 3',5' cyclic nucleotide phosphodiesterase (HSPDE1C1A); PDE1C; phosphodiesterase 1C; phosphodiesterase 1C calmodulin-dependent (70kD); phosphodiesterase 1C, calmodulin-dependent 70kDa
Gene Symbols PDE1C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PDE1C.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5137
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.