Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE2A Antibody (11C6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | PDE2A |
---|---|
Clone | 11C6 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
PDE2A Monoclonal antibody specifically detects PDE2A in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
PDE2A | |
ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cGMP-dependent 3'-5'-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated | |
A synthesized peptide derived from Human PDE2A [UniProt O00408] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
11C6 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5138 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title