Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE2A Antibody (201), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290159AF750
Description
PDE2A Monoclonal specifically detects PDE2A in Human samples. It is validated for ELISA.Specifications
PDE2A | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
201 | |
ELISA | |
cGMP-dependent 3'-5'-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human PDE2A (O00408-1; Glu215-His900). | |
0.1 mL | |
Signal Transduction | |
5138 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction