Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE2A Antibody (OTI3A4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273288H
Description
PDE2A Monoclonal specifically detects PDE2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PDE2A | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PDE2A | |
Full length recombinant protein of human PDE2A (NP_002590) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI3A4 | |
Western Blot, Immunohistochemistry | |
cGMP-dependent 3′-5′-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
5138.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction