Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE2A Monoclonal Antibody (OTI2F1), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA502354

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-3416


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

PDE2A (phosphodiesterase 2A) is a 100 kD cyclic nucleotide phosphodiesterase (PDE) and three isoforms of PDE2A have been reported (PDE2A1, PDE2A2 and PDE2A3) which arise from a single gene by alternative splicing events. PDE2A can degrade both cAMP and cGMP, but degrades cGMP with greater efficacy. PDE2A is expressed in a variety of tissue types, with greatest expression in the brain, where it is localized in the cerebellum and neocortex. PDE2A has been implicated in penile erectile dysfunction and in the regulation of fluid and inflammatory cell transport across the endothelial cell barrier.
Specifications

Specifications

PDE2A
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00408
PDE2A
Full length human recombit protein of human PDE2A produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI2F1
Unconjugated
PDE2A
CGS-PDE, PDE2A1, PED2A4, cGSPDE
Mouse
Affinity Chromatography
RUO
5138
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.