Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE2A Monoclonal Antibody (OTI4D10)
GREENER_CHOICE

Catalog No. PIMA525498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525498 Supplier Invitrogen™ Supplier No. MA525498
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE2A Monoclonal Antibody for Western Blot, ICC/IF, Flow

PDE2A (phosphodiesterase 2A) is a 100 kD cyclic nucleotide phosphodiesterase (PDE) and three isoforms of PDE2A have been reported (PDE2A1, PDE2A2 and PDE2A3) which arise from a single gene by alternative splicing events. PDE2A can degrade both cAMP and cGMP, but degrades cGMP with greater efficacy. PDE2A is expressed in a variety of tissue types, with greatest expression in the brain, where it is localized in the cerebellum and neocortex. PDE2A has been implicated in penile erectile dysfunction and in the regulation of fluid and inflammatory cell transport across the endothelial cell barrier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE2A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4D10
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDE2A
Gene Accession No. O00408
Gene Alias c-GMP stimulated phosphodiesterase; cGMP-dependent 3',5'-cyclic phosphodiesterase; cGMP-stimulated cyclic nucleotide phosphodiesterase; cGMP-stimulated phosphodiesterase 1; cGMP-stimulated phosphodiesterase 2; cGMP-stimulated phosphodiesterase 4; cGSPDE; CGS-PDE; cyclic GMP stimulated phosphodiesterase; cyclic GMP-stimulated phosphodiesterase; cyclic nucleotide phosphodiesterase; cyclic nucleotide phosphodiesterase 2a; Pde2; Pde2a; PDE2A1; Pde2a2; PED2A4; phosphodiesterase 2A; phosphodiesterase 2A, cGMP-stimulated
Gene Symbols PDE2A
Host Species Mouse
Immunogen Full length human recombinant protein of PDE2A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5138
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.