Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571445 Supplier Invitrogen™ Supplier No. PA571445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE2A Polyclonal Antibody for Western Blot

PDE2A (phosphodiesterase 2A) is a 100 kD cyclic nucleotide phosphodiesterase (PDE) and three isoforms of PDE2A have been reported (PDE2A1, PDE2A2 and PDE2A3) which arise from a single gene by alternative splicing events. PDE2A can degrade both cAMP and cGMP, but degrades cGMP with greater efficacy. PDE2A is expressed in a variety of tissue types, with greatest expression in the brain, where it is localized in the cerebellum and neocortex. PDE2A has been implicated in penile erectile dysfunction and in the regulation of fluid and inflammatory cell transport across the endothelial cell barrier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE2A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDE2A
Gene Accession No. O00408
Gene Alias c-GMP stimulated phosphodiesterase; cGMP-dependent 3',5'-cyclic phosphodiesterase; cGMP-stimulated cyclic nucleotide phosphodiesterase; cGMP-stimulated phosphodiesterase 1; cGMP-stimulated phosphodiesterase 2; cGMP-stimulated phosphodiesterase 4; cGSPDE; CGS-PDE; cyclic GMP stimulated phosphodiesterase; cyclic GMP-stimulated phosphodiesterase; cyclic nucleotide phosphodiesterase; cyclic nucleotide phosphodiesterase 2a; Pde2; Pde2a; PDE2A1; Pde2a2; PED2A4; phosphodiesterase 2A; phosphodiesterase 2A, cGMP-stimulated
Gene Symbols PDE2A
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PDE2A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5138
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.