Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE2A1 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502208279
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8279 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8279 Supplier Fabgennix Inc Supplier No. PD2A1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE2A1 Polyclonal antibody specifically detects PDE2A1 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen PDE2A1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PDE2A
Gene Accession No. O00408, Q01062, Q922S4
Gene Alias c-GMP stimulated phosphodiesterase; cGMP-dependent 3',5'-cyclic phosphodiesterase; cGMP-stimulated cyclic nucleotide phosphodiesterase; cGMP-stimulated phosphodiesterase 1; cGMP-stimulated phosphodiesterase 2; cGMP-stimulated phosphodiesterase 4; cGSPDE; CGS-PDE; cyclic GMP stimulated phosphodiesterase; cyclic GMP-stimulated phosphodiesterase; cyclic nucleotide phosphodiesterase; cyclic nucleotide phosphodiesterase 2a; Pde2; Pde2a; PDE2A1; Pde2a2; PED2A4; phosphodiesterase 2A; phosphodiesterase 2A, cGMP-stimulated
Host Species Rabbit
Immunogen Synthetic peptide corresponding to region within amino acids 1-50 on N-terminus of human PDE2A1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207728, 451405, 5138, 81743
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.