Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532971
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532971 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532971 Supplier Invitrogen™ Supplier No. PA532971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Mouse, Rat, Horse, Opossum (100%); Chicken (88%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Cyclic nucleotide phosphodiesterases (PDEs) comprise a complex group of enzymes, and at least 5 major PDE families or classes with distinctive properties have been identified. Members of the cGMP-inhibited cAMP PDE (cGI-PDE) family, such as PDE3A, are characterized by high affinity for cAMP and cGMP and competitive inhibition of their cAMP hydrolytic activity by cGMP and certain positive inotropic agents (Meacci et al., 1992 [PubMed 1315035]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PDE3A
Gene Accession No. Q14432, Q62865, Q9Z0X4
Gene Alias A930022O17Rik; C87899; cAMP phosphodiesterase, myocardial cGMP-inhibited; CGI-PDE; CGI-PDE A; CGI-PDE-A; cGMP-inhibited 3',5'-cyclic phosphodiesterase A; cGMP-inhibited phosphodiesterase; cyclic GMP-inhibited phosphodiesterase A; cyclic nucleotide phosphodiesterase; HTNB; PDE3A; phosphodiesterase 3A; phosphodiesterase 3A, cGMP inhibited; phosphodiesterase 3A, cGMP-inhibited; RNPDE3A
Gene Symbols PDE3A
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human PDE3A.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100034086, 50678, 5139, 54611
Target Species Human, Mouse, Rat, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.