Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115729 Supplier Invitrogen™ Supplier No. PA5115729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PDE3A.

Cyclic nucleotide phosphodiesterases (PDEs) comprise a complex group of enzymes, and at least 5 major PDE families or classes with distinctive properties have been identified. Members of the cGMP-inhibited cAMP PDE (cGI-PDE) family, such as PDE3A, are characterized by high affinity for cAMP and cGMP and competitive inhibition of their cAMP hydrolytic activity by cGMP and certain positive inotropic agents (Meacci et al., 1992 [PubMed 1315035]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE3A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PDE3A
Gene Accession No. Q14432
Gene Alias A930022O17Rik; C87899; cAMP phosphodiesterase, myocardial cGMP-inhibited; CGI-PDE; CGI-PDE A; CGI-PDE-A; cGMP-inhibited 3',5'-cyclic phosphodiesterase A; cGMP-inhibited phosphodiesterase; cyclic GMP-inhibited phosphodiesterase A; cyclic nucleotide phosphodiesterase; HTNB; PDE3A; phosphodiesterase 3A; phosphodiesterase 3A, cGMP inhibited; phosphodiesterase 3A, cGMP-inhibited; RNPDE3A
Gene Symbols PDE3A
Host Species Rabbit
Immunogen A synthesized peptide derived from human PDE3A(Accession Q14432), corresponding to amino acid residues K287-Q337.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5139
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.