Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE3A Rabbit anti-Human, Mouse, Non-human Primate, Rat, Xenopus laevis, Polyclonal, FabGennix

Catalog No. 502208296
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8296 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8296 Supplier Fabgennix Inc Supplier No. PD3A101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE3A Polyclonal antibody specifically detects PDE3A in Human, Mouse, Non-human Primate, Rat, Xenopus laevis samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen PDE3A
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PDE3A
Gene Accession No. Q14432, Q62865, Q9Z0X4
Gene Alias A930022O17Rik; C87899; cAMP phosphodiesterase, myocardial cGMP-inhibited; CGI-PDE; CGI-PDE A; CGI-PDE-A; cGMP-inhibited 3',5'-cyclic phosphodiesterase A; cGMP-inhibited phosphodiesterase; cyclic GMP-inhibited phosphodiesterase A; cyclic nucleotide phosphodiesterase; HTNB; PDE3A; phosphodiesterase 3A; phosphodiesterase 3A, cGMP inhibited; phosphodiesterase 3A, cGMP-inhibited; RNPDE3A
Host Species Rabbit
Immunogen Synthetic peptide selective only for PDE3A variants
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 465329, 50678, 5139, 54611
Target Species Human, Mouse, Non-human Primate, Rat, X. laevis
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.