Learn More
Invitrogen™ PDE3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110087
Description
Immunogen sequence: PLTPFPGFYP CSEIEDPAEK GDRKLNKGLN RNSLPTPQLR RSSGTSGLLP VEQSSRWDRN NGKRPHQEFG ISSQGCYLNG PFNSNLLTIP KQRSSSVSLT HHVGLRRAGV LSSLSPVNSS NHGPVSTGSL TNRSPIEFPD TADFLNKPSV ILQRSLGNAP NTPDFYQQLR NSDSNLCNSC GHQMLKYVST SESDGTDCCS GKSGEEENIF SKESFKLMET QQEEETEKKD SRKLFQEGDK WLTEEAQSEQ QTNIEQEVSL D.
Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.
Specifications
PDE3B | |
Polyclonal | |
Unconjugated | |
PDE3B | |
9830102A01Rik; AI847709; CGIP1; CGI-PDE B; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; HcGIP1; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; PDE3B; phosphodiesterase 3B; phosphodiesterase 3B cGMP inhibited; phosphodiesterase 3B, cGMP-inhibited | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18576, 5140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13370, Q61409 | |
PDE3B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 400-660 of human PDE3B (NP_000913.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.