Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542560 Supplier Invitrogen™ Supplier No. PA542560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SRPEYNFLLH LDHVEFKRFR FLVIEAILAT DLKKHFDFLA EFNAKANDVN Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 79%.

Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE3B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDE3B
Gene Accession No. Q13370
Gene Alias 9830102A01Rik; AI847709; CGIP1; CGI-PDE B; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; HcGIP1; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; PDE3B; phosphodiesterase 3B; phosphodiesterase 3B cGMP inhibited; phosphodiesterase 3B, cGMP-inhibited
Gene Symbols PDE3B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PDE3B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5140
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.