Learn More
PDE3B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302743A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.Specifications
PDE3B | |
Polyclonal | |
Unconjugated | |
PDE3B | |
CGIP1, CGI-PDE B, cGIPDE1, cyclic GMP-inhibited phosphodiesterase B, cyclic nucleotide phosphodiesterase, HcGIP1, phosphodiesterase 3B, cGMP-inhibited | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5140 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q13370 | |
PDE3B | |
Between 1062 and 1112 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.