Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE3B Rabbit anti-Chicken, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502208302
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8302 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8302 Supplier Fabgennix Inc Supplier No. PD3B101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE3B Polyclonal antibody specifically detects PDE3B in Chicken, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PDE3B
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PDE3B
Gene Accession No. Q13370, Q61409, Q63085
Gene Alias 9830102A01Rik; AI847709; CGIP1; CGI-PDE B; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; HcGIP1; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; PDE3B; phosphodiesterase 3B; phosphodiesterase 3B cGMP inhibited; phosphodiesterase 3B, cGMP-inhibited
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1050-2000 on mouse PDE3B protein. Will label all variants of PDE3B.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18576, 29516, 423064, 451041, 5140
Target Species Chicken, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.