Learn More
Invitrogen™ PDE4A Monoclonal Antibody (OTI1C8)

Description
PDE4A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | PDE4A |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1C8 |
| Concentration | 0.92 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Pde4a |
| Gene Accession No. | P27815 |
| Gene Alias | cAMP-specific 3',5'-cyclic phosphodiesterase 4A; cAMP-specific phosphodiesterase PDE4A; cyclic AMP specific phosphodiesterase PDE4A5A; cyclic AMP-specific phosphodiesterase OCPDE4A1A; D9Ertd60e; DPDE2; dunce; LOC100009247; PDE4; PDE46; Pde4a; PHOSA; phosphodiesterase 4A; phosphodiesterase 4A, cAMP specific; phosphodiesterase 4A, cAMP-specific; phosphodiesterase 4A, cAMP-specific (dunce; phosphodiesterase 4A, cAMP-specific (phosphodiesterase E2 dunce homolog, Drosophila); phosphodiesterase E2 dunce homolog, Drosophila; phosphodiesterase isozyme 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.