Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115730 Supplier Invitrogen™ Supplier No. PA5115730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PDE4A.

Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Pde4a
Gene Accession No. O89084, P27815, P54748
Gene Alias cAMP-specific 3',5'-cyclic phosphodiesterase 4A; cAMP-specific phosphodiesterase PDE4A; cyclic AMP specific phosphodiesterase PDE4A5A; cyclic AMP-specific phosphodiesterase OCPDE4A1A; D9Ertd60e; DPDE2; dunce; LOC100009247; PDE4; PDE46; Pde4a; PHOSA; phosphodiesterase 4A; phosphodiesterase 4A, cAMP specific; phosphodiesterase 4A, cAMP-specific; phosphodiesterase 4A, cAMP-specific (dunce; phosphodiesterase 4A, cAMP-specific (phosphodiesterase E2 dunce homolog, Drosophila); phosphodiesterase E2 dunce homolog, Drosophila; phosphodiesterase isozyme 4
Gene Symbols Pde4a
Host Species Rabbit
Immunogen A synthesized peptide derived from human PDE4A(Accession P27815), corresponding to amino acid residues S841-T886.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18577, 25638, 5141
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.