Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE4B Antibody (OTI1D12), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273292PE
Description
PDE4B Monoclonal specifically detects PDE4B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
PDE4B | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
PDE4B | |
Full length recombinant protein of human PDE4B (NP_002591) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI1D12 | |
Flow Cytometry | |
cAMP-specific (phosphodiesterase E4 dunce homolog, cAMP-specific 3′-5′-cyclic phosphodiesterase 4B, cAMP-specific phosphodiesterase-4 B isoform, DKFZp686F2182, Drosophila), dunce-like phosphodiesterase E4, EC 3.1.4, EC 3.1.4.17, MGC126529, PDE32, PDEIVB, phosphodiesterase 4B, cAMP-specific, phosphodiesterase 4B, cAMP-specific (dunce, phosphodiesterase 4B, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E4), phosphodiesterase E4 dunce homolog | |
Mouse | |
Affinity Purified | |
Cancer, Signal Transduction | |
5142.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction