Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4B Monoclonal Antibody (OTI3A10), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA503532

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-4397


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. The cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. This gene encodes a protein that specifically hydrolyzes cAMP. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

PDE4B
Monoclonal
0.73 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q07343
PDE4B
Full length human recombit protein of human PDE4B produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3A10
Unconjugated
PDE4B
DPDE4, PDEIVB
Mouse
Affinity Chromatography
RUO
5142
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.