Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE4B/PDE4C/PDE4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599682 Supplier Invitrogen™ Supplier No. PA599682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PDE4B/C/D.

This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. The encoded protein regulates the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternative splicing and the use of alternative promoters results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4B/PDE4C/PDE4D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PDE4C
Gene Accession No. Q01063, Q07343, Q08493, Q08499, Q3UEI1
Gene Alias 9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4B; cAMP-specific 3',5'-cyclic phosphodiesterase 4C; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D6; cyclic AMP specific phosphodiesterase PDE4D5A; Dpde1; DPDE3; Dpde4; dunce; dunce-like phosphodiesterase E4; E130301F19Rik; HSPDE4D; PDE21; PDE32; PDE43; Pde4b; Pde4c; PDE4D; PDE4DN2; PDEIVB; phosphodiesterase 4B; phosphodiesterase 4B, cAMP specific; phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila); phosphodiesterase 4C; phosphodiesterase 4C, cAMP specific; phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, Drosophila); phosphodiesterase 4D; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); phosphodiesterase E1 dunce homolog; R74983; STRK1; testicular tissue protein Li 136
Gene Symbols PDE4B, PDE4C, PDE4D
Host Species Rabbit
Immunogen A synthesized peptide derived from human PDE4B(Accession Q07343), corresponding to amino acid residues G36-A86.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110385, 18578, 238871, 5142, 5143, 5144
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.