Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4B Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145429
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145429 200 μL
FEPA5145430 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145429 Supplier Thermo Scientific Supplier No. PA5145429

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. The cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. This gene encodes a protein that specifically hydrolyzes cAMP. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4B
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
Gene PDE4B
Gene Accession No. P14646, Q07343
Gene Alias 3',5'-cyclic AMP phosphodiesterase, cAMP-specific 3',5'-cyclic phosphodiesterase 4B, DKFZp686F2182, DPDE4, dunce, dunce-like phosphodiesterase E4, MGC126529, MGC93193, OTTHUMP00000046896, PDE32, PDE4/IVb, PDE4B, Pde4B1, Pde4B2, Pde4B3, Pde4B4, PDEIVB, phosphodiesterase 4B, Phosphodiesterase 4B cAMP-specific (dunce, phosphodiesterase 4B, cAMP specific, phosphodiesterase 4B, cAMP-specific, Phosphodiesterase 4B, cAMP-specific (dunce, phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila), R74983, RP5-876C12.1
Gene Symbols PDE4B
Host Species Rabbit
Immunogen Recombinant Rat PDE4B protein, Lys500-Thr736 (Accession #P14646)
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18578, 24626, 5142
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.