Learn More
Invitrogen™ PDE4C Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLaS3, Raji. Predicted reactivity: Mouse (91%), Rat (92%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PDE4C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.67 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PDE4C |
| Gene Accession No. | Q08493 |
| Gene Alias | cAMP-specific 3',5'-cyclic phosphodiesterase 4C; cAMP-specific 35-cyclic phosphodiesterase 4C; CN4C; DPDE1; dunce; E130301F19Rik; MGC126222; PDE21; Pde4c; phosphodiesterase 4C; phosphodiesterase 4C, cAMP specific; phosphodiesterase 4C, cAMP-specific; phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, Drosophila); phosphodiesterase E1 dunce homolog |
| Gene Symbols | PDE4C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.