Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575546 Supplier Invitrogen™ Supplier No. PA575546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PDE4C
Gene Accession No. P14644, Q08493, Q3UEI1
Gene Alias cAMP-specific 3',5'-cyclic phosphodiesterase 4C; cAMP-specific 35-cyclic phosphodiesterase 4C; CN4C; DPDE1; dunce; E130301F19Rik; MGC126222; PDE21; Pde4c; phosphodiesterase 4C; phosphodiesterase 4C, cAMP specific; phosphodiesterase 4C, cAMP-specific; phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, Drosophila); phosphodiesterase E1 dunce homolog
Gene Symbols PDE4C
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 333-378 of Human PDE4C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110385, 290646, 5143
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.