Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589542 Supplier Invitrogen™ Supplier No. PA589542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKLLQEENCD IFQNLTKKQR QSLRKMVIDI VLATDMSKHM NLLADLKTMV ETKKVTSSGV LLLDNYSDRI QVLQNMVHCA DLSNPTKPLQ LYRQWTDRIM EEFFRQGDRE RERGMEISPM CDKHNASVEK SQVGFIDYIV HPLWETWADL VHPDAQDILD TLEDNREWYQ STIPQSPSPA PDDPEEGRQG QTEKFQFELT LEEDGESDTE KDSGSQVEED TSCSDSKTLC TQDSESTEIP LDEQVEEEAV GEEEESQPEA CVIDDRSPDT; Positive Samples: HT-29, A-549, SW480; Cellular Location: Apical cell membrane, Cytoplasm, Membrane, centrosome, cytoskeleton, microtubule organizing center.

This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pde4d
Gene Accession No. P14270, Q08499
Gene Alias 9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase PDE4D6; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; Dpde3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; HSPDE4D; PDE43; Pde4d; PDE4D3; PDE4D7; PDE4DN2; phosphodiesterase 4D; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); STRK1; testicular tissue protein Li 136
Gene Symbols Pde4d
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 249-518 of human PDE4D (NP_0011841521).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24627, 5144
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.