Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111224 Supplier Invitrogen™ Supplier No. PA5111224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FELTLEEDGE SDTEKDSGSQ VEEDTSCSDS KTLCTQDSES TEIPLDEQVE EEAVG Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 98%.

This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pde4d
Gene Accession No. Q08499
Gene Alias 9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase PDE4D6; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; Dpde3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; HSPDE4D; PDE43; Pde4d; PDE4D3; PDE4D7; PDE4DN2; phosphodiesterase 4D; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); STRK1; testicular tissue protein Li 136
Gene Symbols Pde4d
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PDE4D. Recombinant protein control fragment (Product #RP-96826).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.