Learn More
PDE4D Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302744A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes one of four mammalian counterparts to the fruit fly ′dunce′ gene. The encoded protein has 3′,5′-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.Specifications
PDE4D | |
Polyclonal | |
Unconjugated | |
PDE4D | |
DKFZp686M11213, DPDE3, FLJ97311, HSPDE4D, PDE4DN2, STRK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5144 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q08499 | |
PDE4D | |
Between 709 and 759 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.