Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4D Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208316
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8316 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8316 Supplier Fabgennix Inc Supplier No. PD4401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE4D Polyclonal antibody specifically detects PDE4D in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PDE4D
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Pde4d
Gene Accession No. P14270, Q01063, Q08499
Gene Alias 9630011N22Rik; ACRDYS2; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase PDE4D6; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; Dpde3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; HSPDE4D; PDE43; Pde4d; PDE4D3; PDE4D7; PDE4DN2; phosphodiesterase 4D; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); STRK1; testicular tissue protein Li 136
Host Species Rabbit
Immunogen C-terminal cyclic peptide unique to all PDED subtypes variants.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238871, 24627, 5144
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.