Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550727 Supplier Invitrogen™ Supplier No. PA550727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PDE5A protein.

The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Pde5a
Gene Accession No. O76074
Gene Alias Cgbpde; CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2; phosphodiesterase 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5
Gene Symbols Pde5a
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human phosphodiesterase 5A, cGMP-specific.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8654
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.