Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    PDE5A Antibody, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Goat Polyclonal Antibody
Supplier: Novus Biologicals NB300957
Description
PDE5A Polyclonal specifically detects PDE5A in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
| PDE5A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CGB-PDE, cGMP-binding cGMP-specific 3'-5'-cyclic nucleotide phosphodiesterase, cGMP-binding cGMP-specific phosphodiesterase, cGMP-specific 3'-5'-cyclic phosphodiesterase, cGMP-specific phosphodiesterase type 5A, CN5A, EC 3.1.4, EC 3.1.4.35, PDE5, phosphodiesterase 5A, cGMP-specific, phosphodiesterase isozyme 5 | |
| Goat | |
| 100 kDa | |
| 0.1 mg | |
| Lipid and Metabolism, Signal Transduction | |
| 8654 | |
| Human | |
| IgG | 
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| O76074 | |
| PDE5A | |
| Peptide with sequence C-LINGESGQAKRN corresponding to C-Terminus according to NP_001074.2, NP_246273.2, NP_236914.2, NP_237223. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize reported isoforms NP_001074.2, NP_236914.2 and NP_246273.2. | |
| Store at -20C. Avoid freeze-thaw cycles. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
             
                            