Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6 Beta Rabbit anti-Bovine, Human, Mouse, Porcine, Rat, Polyclonal, FabGennix

Catalog No. 502208396
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8396 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8396 Supplier Fabgennix Inc Supplier No. PD6201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE6 Beta Polyclonal antibody specifically detects PDE6 Beta in Bovine, Human, Mouse, Porcine, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PDE6 Beta
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PDE6B
Gene Accession No. P23439, P23440, P35913
Gene Alias cGMP-phosphodiesterase beta-subunit; CSNB3; CSNBAD2; GMP-PDE beta; Mpb; PDE6B; PDEB; phosphodiesterase 6B; phosphodiesterase 6B, cGMP, rod receptor, beta polypeptide; phosphodiesterase 6B, cGMP-specific, rod, beta; phosphodiesterase, cGMP, rod receptor, beta polypeptide; r; rd; rd1; rd-1; rd10; retinal degeneration 1; rod cGMP-phosphodiesterase beta-subunit; rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit beta; RP40
Host Species Rabbit
Immunogen Synthetic cyclic petide common to all PDE6 Beta variants
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100625223, 18587, 281974, 289878, 5158
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.