Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6a Mouse anti-Human, Unconjugated, Clone: 7A11.1, MilliporeSigma™

Catalog No. MABN2250100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2250-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2250-100 Supplier MilliporeSigma Supplier No. MABN2250100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PDE6a, clone 7A11.1, Cat. No. MABN2250, is a highly specific mouse monoclonal antibody that targets cGMP-PDE alpha and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6a
Applications Western Blot
Classification Monoclonal
Clone 7A11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Rod cGMP-specific 3';5'-cyclic phosphodiesterase subunit alpha;EC: 3.1.4.35;cGMP-PDE alpha;PDE V-B1
Gene Symbols PDE6A;PDEA
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 77 amino acids from the N-terminal region of human Rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha (cGMP-PDE alpha).
Molecular Weight of Antigen ∽100 kDa observed; 99.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000431
Test Specificity Clone 7A11.1 is a mouse monoclonal antibody that specifically detects Rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha (cGMP-PDE alpha) in human retinal tissue. It targets an epitope within 77 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.