Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532974
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532974 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532974 Supplier Invitrogen™ Supplier No. PA532974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster (100%); Dog, Panda, Rabbit (94%); Bovine, Bat, Elephant (89%); Horse, Opossum (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene encodes the cyclic-GMP (cGMP)-specific phosphodiesterase 6A alpha subunit, expressed in cells of the retinal rod outer segment. The phosphodiesterase 6 holoenzyme is a heterotrimer composed of an alpha, beta, and two gamma subunits. cGMP is an important regulator of rod cell membrane current, and its dynamic concentration is established by phosphodiesterase 6A cGMP hydrolysis and guanylate cyclase cGMP synthesis. The protein is a subunit of a key phototransduction enzyme and participates in processes of transmission and amplification of the visual signal. Mutations in this gene have been identified as one cause of autosomal recessive retinitis pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PDE6A
Gene Accession No. P16499, P27664
Gene Alias cGMP phosphodiesterase alpha; cGMP phosphodiesterase alpha subunit; cGMP phosphodiesterase alpha subunit (EC 3.1.4.35); CGPR-A; cyclic GMP phosphodiesterase alpha subunit; GMP phosphodiesterase alpha subunit; GMP-PDE alpha; nmf282; PDE V-B1; PDE6A; PDEA; phosphodiesterase 6A; phosphodiesterase 6A, cGMP-specific, rod, alpha; phosphodiesterase, cyclic GMP (rod receptor), alpha polypeptide; rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha; rod photoreceptor cGMP phosphodiesterase alpha subunit; RP43; unnamed protein product
Gene Symbols PDE6A
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminus of human PDE6A / PDE6 Alpha.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100762641, 225600, 307401, 5145
Target Species Human, Mouse, Rat, Monkey, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.