Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6A Polyclonal Antibody, Invitrogen™

Catalog No. PA1721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1721 Supplier Thermo Scientific Supplier No. PA1721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-721 detects phosphodiesterase 6 alpha’ (PDE6 alpha’) from sheep, human and bovine retinal extract. PA1-721 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽90 kDa protein representing PDE6 alpha’ from sheep and bovine retinal extract. The PA1-721 immunogen is a synthetic peptide corresponding to residues M(16) P Q F A K E Y F D R K L R(29) C of human PDE6 alpha’. This sequence is completely conserved between human, chicken and frog. PA1-721 immunizing peptide (Cat. No. PEP-149) is available for use in neutralization and control experiments.

This gene encodes the cyclic-GMP (cGMP)-specific phosphodiesterase 6A alpha subunit, expressed in cells of the retinal rod outer segment. The phosphodiesterase 6 holoenzyme is a heterotrimer composed of an alpha, beta, and two gamma subunits. cGMP is an important regulator of rod cell membrane current, and its dynamic concentration is established by phosphodiesterase 6A cGMP hydrolysis and guanylate cyclase cGMP synthesis. The protein is a subunit of a key phototransduction enzyme and participates in processes of transmission and amplification of the visual signal. Mutations in this gene have been identified as one cause of autosomal recessive retinitis pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene PDE6A
Gene Accession No. P11541, P16499
Gene Alias Phosphodiesterase 6 Alpha′
Gene Symbols PDE6A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(16) P Q F A K E Y F D R K L R(29) C of human PDE6 alpha’.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281973, 5145, 101122024
Target Species Bovine, Human, Ovine
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.