Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566293 Supplier Invitrogen™ Supplier No. PA566293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLSEEQARSF LDQNPDFARQ YFGKKLSPEN VAAACEDGCP PDCDSLRDLC QVEEST Highest antigen sequence identity to the following orthologs - mouse 81%, rat 79%.

The second messengers cAMP and cGMP are key regulatory molecules that are involved in a wide variety of signal transduction pathways. Levels of cAMP and cGMP are regulated by their rate of synthesis by nucleotide cyclases and by their rate of hydrolysis by cyclic nucleotide phosphodiesterases (PDEs). PDEs form a superfamily of enzymes that catalyze the conversion of 3-prime, 5-prime-cyclic nucleotides to the corresponding nucleoside 5-prime-monophosphates. PDE6 is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha' (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). The catalytic core of the PDE6 system is comprised of alpha'/alpha' homodimers in the cone and alpha/beta heterodimers in the rod. The C-terminus of both the catalytic and inhibitory subunits is modified by methylation, myristyolation and prenylation which have been shown to be critical for proper complex assembly and membrane association.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PDE6B
Gene Accession No. P35913
Gene Alias cGMP-phosphodiesterase beta-subunit; CSNB3; CSNBAD2; GMP-PDE beta; Mpb; PDE6B; PDEB; phosphodiesterase 6B; phosphodiesterase 6B, cGMP, rod receptor, beta polypeptide; phosphodiesterase 6B, cGMP-specific, rod, beta; phosphodiesterase, cGMP, rod receptor, beta polypeptide; r; rd; rd1; rd-1; rd10; retinal degeneration 1; rod cGMP-phosphodiesterase beta-subunit; rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit beta; RP40
Gene Symbols PDE6B
Host Species Rabbit
Immunogen Recombinant Human PDE6B. Recombinant protein control fragment (Product #RP-106974).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5158
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.