Learn More
Invitrogen™ PDE6C Polyclonal Antibody

Description
PDE6C Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | PDE6C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PDE6C |
| Gene Accession No. | P51160, Q91ZQ1 |
| Gene Alias | ACHM5; cGMP phosphodiesterase 6C; COD4; Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'; cpfl1; PDE6C; PDEA2; phosphodiesterase 6C; phosphodiesterase 6C, cGMP specific, cone, alpha prime; phosphodiesterase 6C, cGMP-specific, cone, alpha prime |
| Gene Symbols | PDE6C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.