Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112864
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112864 Supplier Invitrogen™ Supplier No. PA5112864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE6C Polyclonal Antibody for Western Blot, ICC/IF, ELISA

PDE6C is the alpha-prime subunit of cone phosphodiesterase, which is composed of a homodimer of two alpha-prime subunits and 3 smaller proteins of 11, 13, and 15 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PDE6C
Gene Accession No. P51160
Gene Alias ACHM5; cGMP phosphodiesterase 6C; COD4; Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'; cpfl1; PDE6C; PDEA2; phosphodiesterase 6C; phosphodiesterase 6C, cGMP specific, cone, alpha prime; phosphodiesterase 6C, cGMP-specific, cone, alpha prime
Gene Symbols PDE6C
Host Species Rabbit
Immunogen Recombinant Human Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha' protein (285-451AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5146
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.