Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1726
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1726 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1726 Supplier Invitrogen™ Supplier No. PA1726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-726 detects PrBP/delta from bovine, human and sheep samples. PA1-726 has been successfully used in Western blot, ICC/IF and immunohistochemistry procedures. By Western blot, this antibody detects a 17 kDa protein representing PrBP/delta from bovine and sheep retinal/optic nerve extracts as well as HeLa cell lysate. Immunohistochemical staining of PrBP/delta in bovine retinal cross-sections using PA1-726 results in staining of retinal outer segments. PA1-726 immunizing peptides correspond to amino acid residues 1-16 from mouse and human PrBP/delta. This peptide (Cat. # PEP-203) is available for use in neutralization and control experiments.

PrBP/delta, previously known as PDE6/delta, is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). PRBP/delta has been implicated in the transport and membrane targeting of prenylated proteins (including PDE6) from their site of synthesis in the inner segment to their final destination in the outer segment of rods and cones (Norton et. al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6D
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PDE6D
Gene Accession No. O43924, Q95142
Gene Alias AI841218; GMP-PDE delta; JBTS22; PDE6D; PDED; phosphodiesterase 6D; phosphodiesterase 6D, cGMP-specific, rod, delta; PrBP/delta; Protein p17; retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit delta; rod phosphodiesterase delta subunit
Gene Symbols PDE6D
Host Species Rabbit
Immunogen Synthetic Peptides: M(1) S A K D E R A R (D/E) I L R G F K(16) C.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101108369, 281976, 5147
Target Species Human, Bovine, Sheep
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.